missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117926
This item is not returnable.
View return policy
Description
EHD4 Polyclonal Antibody for Western Blot, IP
ATP- and membrane-binding protein that probably controls membrane reorganization/tubulation upon ATP hydrolysis. Plays a role in early endosomal transport. [UniProt].
Specifications
| EHD4 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9H223 | |
| EHD4 | |
| E. coli-derived Human EHD4 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EHD4 | |
| 2210022F10Rik; AI197390; AI846352; AV006278; EH domain containing 4; EH domain containing protein MPAST2; EH domain-containing protein 4; EHD4; EH-domain containing 4; FKSG7; HCA10; HCA11; Hepatocellular carcinoma-associated protein 10/11; hepatocellular carcinoma-associated protein HCA11; mPAST2; ortholog of rat pincher; PAST homolog 2; PAST homolog 4; Past2; PAST4; pincher; pinocytic chaperone | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 30844 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction