missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570659
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
The EHD3 gene encodes the EH-domain containing protein 3, part of the EHD protein family involved in the regulation of endocytic recycling and membrane trafficking. EHD3 plays a crucial role in the endosomal transport system, impacting how receptors and other membrane proteins are recycled back to the cell surface after internalization. This protein aids in maintaining cellular homeostasis by ensuring the efficient sorting and transport of cargo through the endosomal system. EHD3 has been implicated in various cellular functions, including insulin signaling, receptor recycling, and neuronal development, and significantly influences synaptic transmission and plasticity. Alterations in EHD3 expression or function can disrupt endosomal trafficking processes and are linked to conditions such as neurodegenerative diseases and cancer. Research on EHD3 is focused on uncovering its role in membrane dynamics and identifying potential therapeutic strategies for disorders involving dysfunctional endocytic pathways.
Specifications
| EHD3 | |
| Polyclonal | |
| Unconjugated | |
| EHD3 | |
| EH domain containing 3; EH domain-containing protein 3; Ehd2; Ehd3; EH-domain containing 3; EH-domain containing protein 2; PAST homolog 3; PAST3; si:ch211-147c1.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 192249 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8R491 | |
| EHD3 | |
| synthetic peptide directed towards the C-terminal region of rat EHD3. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction