missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549403
This item is not returnable.
View return policy
Description
EHD2 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the EH domain-containing protein family. These proteins are characterized by a C-terminal EF-hand domain, a nucleotide-binding consensus site at the N terminus and a bipartite nuclear localization signal. The encoded protein interacts with the actin cytoskeleton through an N-terminal domain and also binds to an EH domain-binding protein through the C-terminal EH domain. This interaction appears to connect clathrin-dependent endocytosis to actin, suggesting that this gene product participates in the endocytic pathway.
Specifications
| EHD2 | |
| Polyclonal | |
| Unconjugated | |
| Ehd2 | |
| BC027084; C130052H20Rik; EH domain containing 2; EH domain-containing protein 2; Ehd2; EH-domain containing 2; FLJ96617; MGEPS; PAST homolog 2; PAST2; putative eps protein | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 259300, 30846, 361512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q4V8H8, Q8BH64, Q9NZN4 | |
| Ehd2 | |
| KLH conjugated synthetic peptide between 415-449 amino acids from the C-terminal region of human EHD2. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction