missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ EHBP1L1 Recombinant Protein Antigen

Product Code. 18323302
Change view
Click to view available options
:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18323302 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323302 Supplier Novus Biologicals™ Supplier No. NBP317136PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 254102
Molecular Weight (g/mol) M.W. Theoretical: 27 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias DKFZp762C186, EH domain binding protein 1-like 1, EH domain-binding protein 1-like protein 1
Gene Symbol EHBP1L1
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein EHBP1L1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.