missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHBP1L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558804
This item is not returnable.
View return policy
Description
Immunogen sequence: IEVGLLGVLG IETGAAEGAI LGTQEIASRD SGVPGLEADT TGIQVKEVGG SEVPEIATGT AETEILGTQE IASRSSGVPG LESEVAGAQE TEVGGSGISG Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 42%.
Specifications
| EHBP1L1 | |
| Polyclonal | |
| Unconjugated | |
| EHBP1L1 | |
| EH domain binding protein 1 like 1; EH domain binding protein 1-like 1; EH domain-binding protein 1-like protein 1; EHBP1L1; G430002G23Rik; Tangerin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 254102 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N3D4 | |
| EHBP1L1 | |
| Recombinant protein corresponding to Human EHBP1L1. Recombinant protein control fragment (Product #RP-97472). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction