missing translation for 'onlineSavingsMsg'
Learn More

EHBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16877574 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16877574 20 μL 20µL
16867574 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16877574 Supplier Proteintech Supplier No. 176371AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specifications

Antigen EHBP1
Applications Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EHBP1
Gene Accession No. Q69ZW3, Q8NDI1
Gene Alias EH domain binding protein 1, EHBP1, HPC12, KIAA0903, NACSIN
Gene Symbols Ehbp1
Host Species Rabbit
Immunogen EHBP1 Fusion Protein Ag11916
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216565, 23301, 305556
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.