missing translation for 'onlineSavingsMsg'
Learn More

EGTA-AM, MedChemExpress

Product Code. 16481198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481198 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481198 Supplier Medchem Express Supplier No. HYD0973
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress EGTA-​AM is a membrane permeable form of EGTA, can be passively loaded into cells to generate intracellular EGTA; EGTA-​AM is also a Ca2+ chelator with slow chelating dynamics.

EGTA-​AM is a membrane permeable form of EGTA, can be passively loaded into cells to generate intracellular EGTA

Specifications

Chemical Name or Material EGTA-AM
CAS 99590-86-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H40N2O18
Quantity 5 mg
Synonym EGTA Acetoxymethyl ester
Solubility Information DMSO : 50 mg/mL (74.78 mM; Need ultrasonic)
SMILES O=C(OCOC(C)=O)CN(CC(OCOC(C)=O)=O)CCOCCOCCN(CC(OCOC(C)=O)=O)CC(OCOC(C)=O)=O
Molecular Weight (g/mol) 668.6
Formula Weight 668.6
Grade Research
Physical Form Viscous Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.