missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92760-0.1ml
This item is not returnable.
View return policy
Description
EGR4 Polyclonal antibody specifically detects EGR4 in Mouse samples. It is validated for Western Blot
Specifications
| EGR4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| AT133, early growth response 4, early growth response protein 4, EGR-4, NGFIC, NGFI-C, PAT133 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-367 of mouse Egr4 (NP_065621.1). MLHLSDFSGSDALLSKPTEGCAHASPELPRLPARDAPSAAAYPGGDFLSWALSSCGAGGDLTDSCFLEGPAPTPPSGLSYSGSFFIQAVPEHPHDPEALFNLMSGILGLAPFPGPEAAASRSPLDVPFPAGPDALLPDLYSPDLSSAAFPEAFWEAAPSAGAPSQCLFEPQLSPPDVKPGLRAPPASPALDAAASAFKGPYAPWELLSAGVPGNCGSQGSFQTTPEARFSAVGTKVEDLLSISCPAELPGPAARLYQAGAYDTFSLAPGDLGEGTEGLPALLTPPGGEGGSGGEGGEFLAAPPAQLSPLGLRGAATADFSKPLVADLPGGSGVAAPSSPAASFPAAKARRKGRRGGKCSARCFCPRP | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1961 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction