missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103317
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EGR3.
This gene encodes a transcriptional regulator that belongs to the EGR family of C2H2-type zinc-finger proteins. It is an immediate-early growth response gene which is induced by mitogenic stimulation. The protein encoded by this gene participates in the transcriptional regulation of genes in controling biological rhythm. It may also play a role in a wide variety of processes including muscle development, lymphocyte development, endothelial cell growth and migration, and neuronal development. Alternative splicing results in multiple transcript variants encoding distinct isoforms.[provided by RefSeq, Dec 2010].
Specifications
| EGR3 | |
| Polyclonal | |
| Unconjugated | |
| EGR3 | |
| early growth response 3; early growth response protein 3; EGR3; EGR-3; MGC138484; Pilot; zgc:92020; zinc finger protein pilot | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13655, 1960, 25148 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43300, P43301, Q06889 | |
| EGR3 | |
| A synthesized peptide derived from human EGR3(Accession Q06889), corresponding to amino acid residues T124-Q174. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction