Learn More
EGR2 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-92327PE
Product Code. 18525160
  • £418.00 / 0.10mL
Description
EGR2 Polyclonal antibody specifically detects EGR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)