missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR2 Antibody (OTI1B12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70624CL1
This item is not returnable.
View return policy
Description
EGR2 Monoclonal antibody specifically detects EGR2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| EGR2 | |
| Monoclonal | |
| CoraFluor 1 | |
| AT591, CMT1D, CMT4E, early growth response 2, early growth response 2 (Krox-20 homolog, Drosophila), early growth response protein 2, EGR-2, FLJ14547, Krox-20 homolog, Drosophila, KROX-20, Drosophila, homolog (early growth response-2), KROX20DKFZp686J1957, Zinc finger protein Krox-20 | |
| Human recombinant protein fragment corresponding to amino acids 156-476 of human EGR2(NP_000390) produced in E.coli. | |
| 0.1 mL | |
| Chromatin Research, Neurodegeneration, Neuroscience, Transcription Factors and Regulators, Zinc Finger | |
| 1959 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI1B12 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction