missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ EGR2 Antibody Blocking Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Specifications
Specifications
| Gene ID (Entrez) | 1959 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-92327 |
| Gene Alias | AT591, CMT1D, CMT4E, early growth response 2, early growth response 2 (Krox-20 homolog, Drosophila), early growth response protein 2, EGR-2, FLJ14547, Krox-20 homolog, Drosophila, KROX-20, Drosophila, homolog (early growth response-2), KRO x 20 DKFZp686J1957, Zinc finger protein Krox-20 |
| Gene Symbol | EGR2 |
| Quantity | 0.1 mg |
| Research Category | Chromatin Research, Neurodegeneration, Neuroscience, Transcription Factors and Regulators, Zinc Finger |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction