missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR2 Antibody (27C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26605-50ul
This item is not returnable.
View return policy
Description
EGR2 Monoclonal antibody specifically detects EGR2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| EGR2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 27C3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| AT591, CMT1D, CMT4E, early growth response 2, early growth response 2 (Krox-20 homolog, Drosophila), early growth response protein 2, EGR-2, FLJ14547, Krox-20 homolog, Drosophila, KROX-20, Drosophila, homolog (early growth response-2), KROX20DKFZp686J1957, Zinc finger protein Krox-20 | |
| A synthesized peptide derived from Human EGR2 [UniProt P11161] | |
| 50 μL | |
| Chromatin Research, Neurodegeneration, Neuroscience, Transcription Factors and Regulators, Zinc Finger | |
| 1959 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction