Learn More
EGR2 Antibody (27C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26605-100ul
 View more versions of this product
Product Code. 30043208
  • £406.00 / 100µL
Description
EGR2 Monoclonal antibody specifically detects EGR2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EGR2
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Western Blot, Flow Cytometry, ELISA
27C3
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
AT591, CMT1D, CMT4E, early growth response 2, early growth response 2 (Krox-20 homolog, Drosophila), early growth response protein 2, EGR-2, FLJ14547, Krox-20 homolog, Drosophila, KROX-20, Drosophila, homolog (early growth response-2), KROX20DKFZp686J1957, Zinc finger protein Krox-20
A synthesized peptide derived from Human EGR2 [UniProt P11161]
100 μL
Chromatin Research, Neurodegeneration, Neuroscience, Transcription Factors and Regulators, Zinc Finger
1959
Store at -20 to -70C. Avoid freeze-thaw cycles.