missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78775APC
This item is not returnable.
View return policy
Description
EGR1 Polyclonal antibody specifically detects EGR1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, ChIP assay (ChIP)
Specifications
| EGR1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
| AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225Zinc finger protein 225 | |
| The immunogen for this product maps to a region between residue 1 and 50 of mouse Early Growth Response Protein 1 using the numbering given in entry NP_031939.1 (GeneID 13653). | |
| 0.1 mL | |
| Angiogenesis, Cancer, Prostate Cancer, Transcription Factors and Regulators | |
| 1958 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation, ChIP Assay | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction