missing translation for 'onlineSavingsMsg'
Learn More

Eglumegad, MedChemExpress

Product Code. 16487648
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487648

Brand: Medchem Express HY18941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eglumegad (LY354740) is a highly potent and selective group II (mGlu2/3) receptor agonist with IC50s of 5 and 24 nM on transfected human mGlu2 and mGlu3 receptors, respectively.

Eglumegad (LY354740) is a highly potent and selective group II (mGlu2/3) receptor agonist with IC50s of 5 and 24 nM on transfected human mGlu2 and mGlu3 receptors, respectively.

Specifications

Chemical Name or Material Eglumegad
Physical Form Solid
CAS 176199-48-7
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C8H11NO4
For Use With (Application) Neuroscience-Neuromodulation
Synonym LY354740 Eglumetad
Solubility Information H2O : 6.33 mg/mL (34.18 mM; Need ultrasonic and warming)
SMILES OC([C@]1([C@]2([H])[C@@H](C(O)=O)[C@]2([H])CC1)N)=O
Molecular Weight (g/mol) 185.18
Formula Weight 185.18
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.