missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN3/PHD3 Rabbit anti-Human, Mouse, Rat, Clone: 4S7S3, Novus Biologicals™
Rabbit Monoclonal Antibody
£400.00
Specifications
| Antigen | EGLN3/PHD3 |
|---|---|
| Clone | 4S7S3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18344295
|
Novus Biologicals
NBP3-15313-20UL |
20 μg |
N/A
|
N/A | |||||
|
18388013
|
Bio-Techne
NBP3-15313-100UL |
100 μg |
£400.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGLN3/PHD3 Monoclonal antibody specifically detects EGLN3/PHD3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| EGLN3/PHD3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Chromatin Research, Hypoxia | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 112399 | |
| IgG | |
| Affinity purified |
| 4S7S3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-239 of human EGLN3/PHD3 (Q9H6Z9). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title