missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN3/PHD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | EGLN3/PHD3 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18282653
|
Novus Biologicals
NBP2-57377 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657238
|
Novus Biologicals
NBP2-57377-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGLN3/PHD3 Polyclonal specifically detects EGLN3/PHD3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| EGLN3/PHD3 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Chromatin Research, Hypoxia | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 112399 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YFDAEERAEAKKKFRNLTRKTESALTED | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
| EGLN3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title