missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN3/PHD3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 46 publications
£207.00 - £463.00
Specifications
| Antigen | EGLN3/PHD3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24367580), Immunohistochemistry 1:250-1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:250-1:1000, Electron Microscopy reported in scientific literature (PMID 17003483), Mass Spectrometry, KnockDown Validated |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18710024
|
Novus Biologicals
NB100-139SS |
0.025 mL |
£207.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776283
|
Novus Biologicals
NB100-139 |
£463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
EGLN3/PHD3 Polyclonal specifically detects EGLN3/PHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy, Mass Spectrometry, Knockdown Validated.Specifications
| EGLN3/PHD3 | |
| Western Blot 1:1000-1:2000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24367580), Immunohistochemistry 1:250-1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:250-1:1000, Electron Microscopy reported in scientific literature (PMID 17003483), Mass Spectrometry, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Hypoxia | |
| PBS with 0.05% Sodium Azide | |
| EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
| EGLN3 | |
| IgG | |
| Affinity Purified | |
| Human and rat. Mouse reactivity reported in scientific literature (PMID: 24037093). |
| 1 mg/mL | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Q9H6Z9 | |
| 112399 | |
| Synthetic peptide corresponding to residues between 50-100 of human PHD3/HIF Prolyl Hydroxylase 3 using the numbering given in entry NP_071356.1 (GeneID 112399). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 28 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title