missing translation for 'onlineSavingsMsg'
Learn More

EGLN3/PHD3 Antibody (700210) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30737893 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737893 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737893 Supplier Novus Biologicals Supplier No. FAB6954MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EGLN3/PHD3 Monoclonal antibody specifically detects EGLN3/PHD3 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGLN3/PHD3
Applications Immunocytochemistry
Classification Monoclonal
Clone 700210
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3
Host Species Mouse
Immunogen E. coli-derived recombinant human EGLN3/PHD3, Pro2-Asp239, Accession # Q9H6Z9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 112399
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.