missing translation for 'onlineSavingsMsg'
Learn More

EGLN2/PHD1 Rabbit anti-Human, Mouse, Rat, Clone: 2G9M7, Novus Biologicals™

Product Code. 18004964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18004964 20 μg 20µL
18388596 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18004964 Supplier Novus Biologicals Supplier No. NBP31626520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EGLN2/PHD1 Monoclonal antibody specifically detects EGLN2/PHD1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGLN2/PHD1
Applications Western Blot
Classification Monoclonal
Clone 2G9M7
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DKFZp434E026, EC 1.14.11, EC 1.14.11.-, egl nine homolog 2, egl nine homolog 2 (C. elegans), EIT6, Estrogen-induced tag 6, FLJ95603, HIF-PH1, HIFPH1 EGL nine (C.elegans) homolog 2, HIF-prolyl hydroxylase 1, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 1, PHD1, Prolyl hydroxylase domain-containing protein 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human EGLN2/PHD1 (Q96KS0). MDSPCQPQPLSQALPQLPGSSSEPLEPEPGRARMGVESYLPCPLLPSYHCPGVPSEASAGSGTPRATATSTTASPLRDGFGGQDGGELRPLQSEGAAALV
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 112398
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.