missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN2/PHD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £494.00
Specifications
| Antigen | EGLN2/PHD1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663931
|
Novus Biologicals
NBP2-76551-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18297299
|
Novus Biologicals
NBP2-76551 |
100 μL |
£494.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGLN2/PHD1 Polyclonal specifically detects EGLN2/PHD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| EGLN2/PHD1 | |
| Polyclonal | |
| Rabbit | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| DKFZp434E026, EC 1.14.11, EC 1.14.11.-, egl nine homolog 2, egl nine homolog 2 (C. elegans), EIT6, Estrogen-induced tag 6, FLJ95603, HIF-PH1, HIFPH1 EGL nine (C.elegans) homolog 2, HIF-prolyl hydroxylase 1, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 1, PHD1, Prolyl hydroxylase domain-containing protein 1 | |
| EGLN2 | |
| IgG | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Human | |
| Q96KS0 | |
| 112398 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PLSQALPQLPGSSSEPLEPEPGRARMGVESYLPCPLLPSYHCPGVPSEASAGSGTPRATATSTTASPLRDGFGGQDGGE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title