missing translation for 'onlineSavingsMsg'
Learn More

EGLN2/PHD1 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18402339
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402339 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402339 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP305472UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EGLN2/PHD1 Polyclonal antibody specifically detects EGLN2/PHD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGLN2/PHD1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias DKFZp434E026, EC 1.14.11, EC 1.14.11.-, egl nine homolog 2, egl nine homolog 2 (C. elegans), EIT6, Estrogen-induced tag 6, FLJ95603, HIF-PH1, HIFPH1 EGL nine (C.elegans) homolog 2, HIF-prolyl hydroxylase 1, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 1, PHD1, Prolyl hydroxylase domain-containing protein 1
Host Species Rabbit
Immunogen Partial synthetic peptide made to an N-terminal portion of the human EGLN2/ PHD1 protein (between amino acids 10-75) [UniProt Q96KS0]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 112398
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.