missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN2/PHD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92974-0.02ml
This item is not returnable.
View return policy
Description
EGLN2/PHD1 Polyclonal antibody specifically detects EGLN2/PHD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| EGLN2/PHD1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:20-1:50 | |
| DKFZp434E026, EC 1.14.11, EC 1.14.11.-, egl nine homolog 2, egl nine homolog 2 (C. elegans), EIT6, Estrogen-induced tag 6, FLJ95603, HIF-PH1, HIFPH1 EGL nine (C.elegans) homolog 2, HIF-prolyl hydroxylase 1, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 1, PHD1, Prolyl hydroxylase domain-containing protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human EGLN2/PHD1 (NP_444274.1). EVEALKRGGRLRDGQ | |
| 0.02 mL | |
| Cancer, Hypoxia, Signal Transduction | |
| 112398 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction