missing translation for 'onlineSavingsMsg'
Learn More

EGLN1/PHD2 Antibody (2445B), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18408107 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408107 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408107 Supplier Novus Biologicals Supplier No. NBP276810JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EGLN1/PHD2 Monoclonal antibody specifically detects EGLN1/PHD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGLN1/PHD2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2445B
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias C1orf12, EC 1.14.11.29, EC:1.14.11.291, ECYT3, egl nine homolog 1, egl nine-like protein 1, EGLN1, HALAH, HIF prolyl hydroxylase 2, HIFPH2, HIF-PH2, HIF-prolyl hydroxylase 2, HPH2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2, PNAS-118, PNAS-137, Prolyl hydroxylase domain-containing protein 2, SM20, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6
Host Species Rabbit
Immunogen This EGLN1/PHD2 antibody was developed against a partial synthetic peptide from the N terminal portion of the human EGLN1/PHD2 protein.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Chromatin Research, Hypoxia, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 54583
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.