missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN1/PHD2 Antibody (2445B) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£137.00 - £328.00
Specifications
| Antigen | EGLN1/PHD2 |
|---|---|
| Clone | 2445B |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 μ/mL, Flow Cytometry 0.25 ug, Immunohistochemistry 5 - 20 μ/mL, Immunocytochemistry/Immunofluorescence 5 - 20 μ/mL, Immunohistochemistry-Paraffin 5 - 20 μ/mL, Flow (Intracellular) 0.25 ug |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18310151
|
Novus Biologicals
NBP2-76810SS |
0.025 mg |
£137.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18074526
|
Novus Biologicals
NBP2-76810 |
0.1 mg |
£328.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
| EGLN1/PHD2 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Human | |
| C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
| EGLN1 | |
| IgG | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 2445B | |
| Western Blot 1 - 2 μ/mL, Flow Cytometry 0.25 ug, Immunohistochemistry 5 - 20 μ/mL, Immunocytochemistry/Immunofluorescence 5 - 20 μ/mL, Immunohistochemistry-Paraffin 5 - 20 μ/mL, Flow (Intracellular) 0.25 ug | |
| Monoclonal | |
| Rabbit | |
| PBS with 0.02% sodium azide | |
| 54583 | |
| Partial synthetic peptide from the N terminal portion of the human EGLN1/PHD2 protein | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title