Learn More
Neogen Egg White Protein ELISA Kit, E96EGG
Click to view available options
Quantity:
96 Wells
Unit Size:
1 set
Description
- Designed to detect both processed and unprocessed target allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
- A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifications
Specifications
| Product Type | Test |
| Quantity | 96 Wells |
| Test Time | 50 Minutes |