missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX EGFR Protein, Active

Product Code. 17380304
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17380304

Brand: Eurofins DiscoverX 14531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal GST-tagged human EGFR kinase domain, amino acids 696-end, for use in Enzyme Assays.

Receptors that are tyrosine protein kinases include those receptors for epidermal growth factor (EGF), platelet-derived growth factor (PDGF), insulin, and certain lymphokines. Receptor tyrosine kinases are cell surface transmembrane proteins responsible for intracellular signal transduction. They are expressed in many cell types and, after activation by growth factor binding, trigger a series of intracellular pathways leading to a wide variety of responses; e.g. differentiation, proliferation, migration and invasion, angiogenesis, survival. EGF receptor agonism may facilitate wound healing, enhance viral infections, topical inflammation and asthma as well as tumor cell proliferation and metastasis.

GenBank X00588

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 86 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥54%
Protein EGFR, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.