missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGFR, p Tyr1092 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£195.00 - £321.00
Specifications
| Antigen | EGFR (p Tyr1092) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18250362
|
Novus Biologicals
NB100-81894-0.025ML |
0.025 mL |
£195.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18262300
|
Novus Biologicals
NB100-81894 |
0.05 mL |
£321.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGFR Polyclonal specifically detects EGFR in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EGFR (p Tyr1092) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| P00533 | |
| 1956 | |
| IgG | |
| Affinity Purified | |
| 175 kDa |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| EGFR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title