missing translation for 'onlineSavingsMsg'
Learn More

EGFr Mouse Monoclonal Antibody (clone 31G7), Concentrate

Product Code. 10631804
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10631804 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10631804 Supplier Invitrogen Supplier No. 280005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

EGFr Mouse Monoclonal Antibody (clone 31G7), Concentrate

This monoclonal antibody reacts with the extracellular domain of epidermal growth factor receptor (170 kDa). Clone 31G7 is useful in detecting the majority of squamous cell carcinomas that arise from both squamous epithelium and metaplastic squamous epithelium. Positive tissue: Breast carcinoma.

Clone/PAD: 31G7

Immunogen: EGFr derived from A-431 cells

Specificity: EGFr. No cross-reactivity observed with c-erbB-2.

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Not required , Enzyme Digestion: Required (Digest-All™ 3/Pepsin)

Expected Staining Pattern: Cell membrane and cytoplasm

“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFR
Applications Anatomical Pathology, Immunohistochemistry, Immunohistochemistry (Clinical - Staining and Detection), Immunohistochemistry (Clinical), Immunohistochemistry (FFPE)
Classification Monoclonal
Clone 31G7
Conjugate Unconjugated
Dilution Concentrate antibody
Host Species Mouse
Immunogen EGFr derived from A-431 cells
Positive Control Breast carcinoma
Quantity 2 mL
Regulatory Status RUO, IVD/CE/Class 2
Primary or Secondary Primary
Gene ID (Entrez) 1956
Packaging 1 Each
Test Specificity EGFR
Includes 0.1% Sodium azide
Target Species Human
Content And Storage Store at 2 - 8 °C.
Form Concentrated
Isotype IgG1
Show More Show Less

For In Vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.