Learn More
Invitrogen™ Human EGFR (ErbB1) [T790M, L858R], GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic
Specifications
Specifications
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% polysorbate 20, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Molecular Weight (g/mol) | 90.6 kDa |
| Name | Human EGFR (ErbB1) [T790M, L858R], GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Common Name | EGFR (T790M, L858R Mutant) |
| KinaseFamily | EGF Receptor (ErbB) Kinase Family |
| Protein Length | 668-1210; T790M, L858R |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.