missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EGFR (ErbB1), GST Tag Recombinant Protein
Human EGFR (ErbB1), GST Tag Recombinant Protein
£433.00 - £6900.00
Specifications
Accession Number | NP_005219.2 |
---|---|
Gene Alias | EGFR (ErbB1) |
KinaseFamily | EGF Receptor (ErbB) Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10404233
|
Invitrogen™
PV4190 |
1mg |
£6900.00
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418553
|
Invitrogen™
PR7295B |
100μg |
£1624.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11593597
|
Invitrogen™
PV3872 |
10 μg |
£433.00
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics.
Specifications
NP_005219.2 | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Baculovirus |
EGFR (ErbB1) | |
Human | |
GST Tag |