missing translation for 'onlineSavingsMsg'
Learn More

EGFR Antibody (C225 (Cetuximab)), DyLight 488, Novus Biologicals™

Product Code. 18396090 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396090 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396090 Supplier Novus Biologicals Supplier No. NBP275903G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFR
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone C225 (Cetuximab)
Conjugate DyLight 488
Dilution Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Gene Symbols EGFR
Host Species Human
Immunogen The epidermal growth factor receptor is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 1956.0
Test Specificity Recognises human EGFR.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.