missing translation for 'onlineSavingsMsg'
Learn More

EGFLAM Antibody (5A5), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43018

Product Code. 30279657

  • £336.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EGFLAM Monoclonal antibody specifically detects EGFLAM in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EGFLAM
Monoclonal
Unconjugated
PBS, pH 7.3, 50% glycerol
Mouse
Protein A or G purified
RUO
133584
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
ELISA, Western Blot
5A5
Western Blot 1:500 - 1:2000, ELISA
AGRINLAGRNLFLJ39155, Agrin-like protein, EGF-like, fibronectin type III and laminin G domains, EGF-like, fibronectin type-III and laminin G-like domain-containing protein, pikachurin
EGF-like, fibronectin type III and laminin G domains - (Uniprot# Q63HQ2)
100 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.