missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGFL6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551642
This item is not returnable.
View return policy
Description
Immunogen sequence: CSFNHGICDW KQDREDDFDW NPADRDNAIG FYMAVPALAG HKKDIGRLKL LLPDLQPQSN FCLLFDYRLA GDKVGKLRVF VKNSNNALAW EKTTSEDEKW KTGKIQLYQG TDATKSIIFE AERGKGKTGE IAVDGVLLVS GLCPD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
The EGFL6 gene encodes epidermal growth factor-like domain-containing protein 6, a member of the EGF-like repeat superfamily known for its roles in development and angiogenesis. EGFL6 is expressed at higher levels during embryonic development and has notable expression in various malignant tumors, highlighting its potential involvement in cancer progression. This secreted protein is pivotal in promoting tumor angiogenesis, influencing cancer cell migration and proliferation without affecting wound healing, making it a significant factor in tumor growth and metastasis. EGFL6 is also expressed in adipose tissues, and its role in adipose differentiation suggests it might link obesity to endometrial cancer risks. The protein impacts cellular activities such as cell adhesion, migration, and proliferation, contributing fundamentally to tumor dynamics. Consequently, EGFL6 is considered an attractive target for antibody-based therapies, with research evaluating its utility in cancer intervention strategies.
Specifications
| EGFL6 | |
| Polyclonal | |
| Unconjugated | |
| EGFL6 | |
| EGF like domain multiple 6; EGF repeat-containing protein 6; EGFL6; EGF-like protein 6; EGF-like-domain, multiple 6; Epidermal growth factor-like protein 6; Maeg; MAM and EGF domain containing; MAM and EGF domains-containing gene protein; PP648; Protein W80; UNQ281/PRO320; W80 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25975 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUX8 | |
| EGFL6 | |
| Recombinant protein corresponding to Human EGFL6. Recombinant protein control fragment (Product #RP-94750). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction