missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGFL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106685
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MEGF9.
MEGF9 is a protein coding gene.
Specifications
| EGFL5 | |
| Polyclonal | |
| Unconjugated | |
| MEGF9 | |
| 4933405H16Rik; 9630025C07; EGFL5; EGF-like protein 5; EGF-like-domain, multiple 5; Epidermal growth factor-like protein 5; KIAA0818; Megf9; mKIAA0818; multiple EGF like domains 9; multiple EGF-like domains protein 9; multiple EGF-like-domains 9; multiple epidermal growth factor-like domains protein 9; UNQ671/PRO1305 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1955, 230316, 313270 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BH27, Q9H1U4 | |
| MEGF9 | |
| A synthesized peptide derived from human MEGF9(Accession Q9H1U4), corresponding to amino acid residues C383-W433. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction