missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGFL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA537453
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody detects endogenous protein at a molecular weight of 255 kDa. Purity is >95% by SDS-PAGE.
EGFL4 (epidermal growth factor-like protein 4), also known as MEGF8 (multiple epidermal growth factor-like domains protein 8) or C19orf49, is a 2,845 amino acid single-pass type I membrane protein. EGFL4 contains 2 CUB domains, 5 EGF-like domains, 12 Kelch repeats, 4 laminin EGF-like domains and 7 PSI domains. Existing as two alternatively spliced isoforms, the gene encoding EGFL4 maps to human chromosome 19q13.2. Chromosome 19 consists of approximately 63 million bases, makes up over 2% of human genomic DNA, and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin superfamily members including the killer cell and leukocyte Ig-like receptors.
Especificaciones
| EGFL4 | |
| Polyclonal | |
| Unconjugated | |
| MEGF8 | |
| AW049492; b2b1702Clo; b2b288Clo; C19orf49; CRPT2; EGFL4; EGF-like domain-containing protein 4; EGF-like protein 4; EGF-like-domain, multiple 4; Epidermal growth factor-like protein 4; HBV pre-s2 binding protein 1; HBV pre-S2-binding protein 1; hepatitis B virus pre-S2-binding protein 1; KIAA0817; m687Ddg; Megf8; mKIAA0817; multiple EGF like domains 8; multiple EGF-like domain protein 4; multiple EGF-like domains protein 8; multiple EGF-like-domains 8; multiple epidermal growth factor-like domains protein 8; SBP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114029, 1954, 269878 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P60882, Q7Z7M0, Q9QYP0 | |
| MEGF8 | |
| Synthetic peptide corresponding to amino acids 1245-1296 of human EGFL4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido