Learn More
PeproTech EGF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P277BT-500UG
Description
AA Sequence of recombinant protein: MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat EGF. Anti-Rat EGF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat EGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat EGF (500-P277) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat EGF. Western Blot: To detect Rat EGF by Western Blot analysis this antibody can be used at a concentration of 0.5-1.0 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat EGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P277BT-1MG will be provided as 2 x 500 μg
EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
Specifications
| EGF | |
| Polyclonal | |
| Biotin | |
| EGF | |
| AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25313 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P07522 | |
| EGF | |
| E.coli-derived Recombinant Rat EGF. | |
| 500 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.