missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGF Monoclonal Antibody (9A1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ M807
This item is not returnable.
View return policy
Description
The M807 anti-EGF antibody has successfully been paired as the detection antibody in a sandwich ELISA with coating antibody MA805 (Clone 1H11). Typical dilutions for sandwich ELISA: Coat = 1-3 μg/mL and Detection = 0.125-0.5 μg/mL.
EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
Specifications
| EGF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P01133 | |
| EGF | |
| Recombinant Human EGF. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 9A1 | |
| Unconjugated | |
| EGF | |
| AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone | |
| Mouse | |
| Protein A | |
| RUO | |
| 1950 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction