missing translation for 'onlineSavingsMsg'
Learn More

EGF Antibody (10825) [DyLight 350], Novus Biologicals™

Product Code. 30734362 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734362 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734362 Supplier Novus Biologicals Supplier No. FAB236D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGF
Applications Immunohistochemistry, Western Blot, Neutralization
Classification Monoclonal
Clone 10825
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG
Host Species Mouse
Immunogen E. coli-derived recombinant human EGF
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1950
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.