missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EGCG

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
β-secretase (BACE1) inhibitor (IC50 = 1.6 μM); inhibits amyloid assembly. Inhibits the formation of amyloid fibrils by proteins such as Aβ, α-synuclein and huntingtin. Reduces Aβ42-induced cell death in three different cell types. Also inhibits glutamate dehydrogenase. Inhibits cell growth and selectively induces apoptosis in A431 and human pancreatic cancer cells. Also a DNMT1 inhibitor.
Specifications
Specifications
| CAS | 989-51-5 |
| Quantity | 50 mg |
| Molecular Formula | C22H18O11 |
| Formula Weight | 458.37 |
| Purity | >99% |
| Molecular Weight (g/mol) | 458.375 |
| Chemical Name or Material | EGCG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction