missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eg5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23333-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.Specifications
| Eg5 | |
| Polyclonal | |
| Unconjugated | |
| KIF11 | |
| EG5, HKSP, KIF11, kinesin family member 11, Kinesin like protein 1, Kinesin like protein KIF11, KNSL1, TR interacting protein 5, TRIP 5, TRIP5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3832 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52732 | |
| KIF11 | |
| Eg5 Fusion Protein Ag18695 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction