missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eg5 Rabbit anti-Human, Clone: 6G8V8, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16458-20UL
This item is not returnable.
View return policy
Description
Eg5 Monoclonal antibody specifically detects Eg5 in Human samples. It is validated for Western Blot
Specifications
| Eg5 | |
| Monoclonal | |
| Unconjugated | |
| Eg5, HKSP, kinesin family member 11, kinesin-like 1, Kinesin-like protein 1, kinesin-like protein KIF11, Kinesin-like spindle protein HKSP, Kinesin-related motor protein Eg5, KNSL1TRIP-5, thyroid receptor interacting protein 5, Thyroid receptor-interacting protein 5, TR-interacting protein 5, TRIP5EG5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 900-1056 of human Eg5 (P52732). | |
| 20 μg | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 3832 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 6G8V8 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction