missing translation for 'onlineSavingsMsg'
Learn More

Eg5 Inhibitor VII, TRC

Product Code. 30388147
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388147 Supplier Toronto Research Chemicals Supplier No. TRCE44005350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eg5 Inhibitor VII
CAS 912953-25-4
Molecular Formula C16 H12 F3 N O
Quantity 50 mg
Synonym 3,4-Dihydro-6-[4-(trifluoromethyl)phenyl]-2(1H)-quinolinone 6-(4-Trifluoromethylphenyl)-3,4-dihydro-1H-quinolin-2-one 6-[4-(Trifluoromethyl)phenyl]-3,4-dihydroquinolin-2(1H)-one GSK 1
InChI Formula InChI=1S/C16H12F3NO/c17-16(18,19)13-5-1-10(2-6-13)11-3-7-14-12(9-11)4-8-15(21)20-14/h1-3,5-7,9H,4,8H2,(H,20,21)
SMILES FC(F)(F)c1ccc(cc1)c2ccc3NC(=O)CCc3c2
IUPAC Name 6-[4-(trifluoromethyl)phenyl]-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol) 291.268
Formula Weight 291.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.