missing translation for 'onlineSavingsMsg'
Learn More

Eg5 Antibody, Novus Biologicals™

Product Code. 18701414 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18701414 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18701414 Supplier Novus Biologicals Supplier No. NB500181

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 18 publications

Eg5 Polyclonal specifically detects Eg5 in Human, Mouse, Rat, Porcine, Drosophila, Mammal, Plant samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoblotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eg5
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500, Immunoblotting reported in scientific literature (PMID 28392145)
Formulation Whole antisera with No Preservative
Gene Accession No. P52732
Gene Alias Eg5, HKSP, kinesin family member 11, kinesin-like 1, Kinesin-like protein 1, kinesin-like protein KIF11, Kinesin-like spindle protein HKSP, Kinesin-related motor protein Eg5, KNSL1TRIP-5, thyroid receptor interacting protein 5, Thyroid receptor-interacting protein 5, TR-interacting protein 5, TRIP5EG5
Gene Symbols KIF11
Host Species Rabbit
Immunogen A recombinant segment of the coiled-coil domain of human Eg5 [Uniprot: P52732]
Molecular Weight of Antigen 125 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Cytoskeleton Markers, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3832
Test Specificity Human, rat and porcine. Predicted to react with most mammalian species. Immunogen sequence has 98% identity to primate. Drosophila reactivity reported in scientific literature (PMID: 23888285). Silvetia compressa reactivity reported in scientific literature (PMID: 19527496).
Target Species Human, Mouse, Rat, Pig, Drosophila, Mammalia, Plant
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.