missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eg5 Antibody (1G11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26182-50ul
This item is not returnable.
View return policy
Description
Eg5 Monoclonal antibody specifically detects Eg5 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Eg5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1G11 | |
| Western Blot 1:500-1:5000, ELISA | |
| Eg5, HKSP, kinesin family member 11, kinesin-like 1, Kinesin-like protein 1, kinesin-like protein KIF11, Kinesin-like spindle protein HKSP, Kinesin-related motor protein Eg5, KNSL1TRIP-5, thyroid receptor interacting protein 5, Thyroid receptor-interacting protein 5, TR-interacting protein 5, TRIP5EG5 | |
| A synthesized peptide derived from Human Eg5 [UniProt P52732] | |
| 50 μL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3832 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction