missing translation for 'onlineSavingsMsg'
Learn More

EG-VEGF/PK1 Antibody (SR1180), Novus Biologicals™

Product Code. 18630165 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18630165 100 μg 100µL
18613725 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18630165 Supplier Novus Biologicals Supplier No. NBP322375100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

EG-VEGF/PK1 Monoclonal antibody specifically detects EG-VEGF/PK1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EG-VEGF/PK1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SR1180
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1
Host Species Rabbit
Immunogen A synthesized peptide derived from human EG-VEGF/PK1 (Uniprot #: P58294)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 84432
Target Species Human, Mouse
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.