missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EG-VEGF/PK1 Antibody (SR1180), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22375-50ul
This item is not returnable.
View return policy
Description
EG-VEGF/PK1 Monoclonal antibody specifically detects EG-VEGF/PK1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| EG-VEGF/PK1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1180 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1 | |
| A synthesized peptide derived from human EG-VEGF/PK1 (Uniprot #: P58294) | |
| 50 μg | |
| Cell Cycle and Replication | |
| 84432 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction