missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EFTUD2 Recombinant Rabbit Monoclonal Antibody (2Z1K3)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557936

 View more versions of this product

Product Code. 30283330

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MDTDLYDEFG NYIGPELDSD EDDDELGRET KDLDEMDDDD DDDDVGDHDD DHPGMEVVLH EDKKYYPTAE EVYGPEVETI VQEEDTQPLT EPIIKPVKTK KFTLMEQTLP VTVYEMDFLA DLMDNSELIR NVTLCGHLHH GKTCFVDCLI EQTHPEIRKR YDQDLCYTDI LFTEQERGVG IKSTPVTVVL PDTKGKSYLF NIMDT.

EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EFTUD2
Recombinant Monoclonal
2.51 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
O08810, Q15029
EFTUD2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-205 of human EFTUD2 (NP_004238.3).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
2Z1K3
Unconjugated
EFTUD2
116 kDa U5 small nuclear ribonucleoprotein component; 116kDa; Eftud2; elongation factor Tu GTP binding domain containing 2; elongation factor Tu GTP-binding domain-containing protein 2; hSNU114; KIAA0031; MFDGA; MFDM; SNRNP116; Snrp116; Snu114; SNU114 homolog; U5 small nuclear ribonucleoprotein 116 kDa; U5 snRNP-specific protein, 116 kDa; U5-116 kDa; U5-116KD
Rabbit
Protein A
RUO
20624, 287739, 9343
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.