missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EFTUD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554554
This item is not returnable.
View return policy
Description
Immunogen sequence: DDGVQFHAFG RVLSGTIHAG QPVKVLGENY TLEDEEDSQI CTVGRLWISV ARYHIEVNRV PAGNWVLIEG VDQPIVKTAT ITEPRGNEEA QIFRPLKFNT TSVIKIAVEP V Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.
Specifications
| EFTUD2 | |
| Polyclonal | |
| Unconjugated | |
| EFTUD2 | |
| 116 kDa U5 small nuclear ribonucleoprotein component; 116kDa; Eftud2; elongation factor Tu GTP binding domain containing 2; elongation factor Tu GTP-binding domain-containing protein 2; hSNU114; KIAA0031; MFDGA; MFDM; SNRNP116; Snrp116; Snu114; SNU114 homolog; U5 small nuclear ribonucleoprotein 116 kDa; U5 snRNP-specific protein, 116 kDa; U5-116 kDa; U5-116KD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9343 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15029 | |
| EFTUD2 | |
| Recombinant protein corresponding to Human EFTUD2. Recombinant protein control fragment (Product #RP-93217). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction